2,3-Dichloropyridine ≥98% for synthesis

Supplier: Matrix Scientific
Warning

011138-25G 011138-100G
101799-980EA 23.08 USD
101799-980 101799-982
2,3-Dichloropyridine ≥98% for synthesis
2,3-Dichloropyridine

Hazard code(s): Irritant

Formula: C₅H₃Cl₂N
Boiling Pt: 202.5 - 203.5 °C (1013 hPa)
Melting Pt: 70 - 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00006229
CAS Number: 2402-77-9
EINECS: 219-281-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 65-67°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR