2-Fluoro-6-nitrophenol ≥98%

Supplier: Matrix Scientific
Warning

011134-5G 011134-25G 011134-100G
101799-966EA 27.69 USD
101799-966 101799-968 101799-970
2-Fluoro-6-nitrophenol ≥98%
2-Fluoro-6-nitrophenol

Hazard code(s): Irritant

Formula: C₆H₄FNO₃
Melting Pt: 91 - 92 °C
MDL Number: MFCD00042446
CAS Number: 1526-17-6
EINECS: 216-199-4

Order Now

Specification Test Results

Melting point 92-94°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR