5-Amino-2-fluoropyridine ≥95%

Supplier: Matrix Scientific

011131-5G 011131-25G
101799-958EA 52.31 USD
101799-958 101799-960
5-Amino-2-fluoropyridine ≥95%
5-Amino-2-fluoropyridine

Hazard code(s): Toxic

Formula: C₅H₅FN₂
Boiling Pt: 88…91 °C
MDL Number: MFCD01632180
CAS Number: 1827-27-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR