2-Chloro-3-hydroxypyridine ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2-Chloro-3-pyridinol

011130-25G 011130-100G
101799-954EA 35.38 USD
101799-954 102832-828
2-Chloro-3-hydroxypyridine ≥98%
2-Chloro-3-hydroxypyridine

Hazard code(s): Irritant

Formula: C₅H₄ClNO
MW: 129.55 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 170…172 °C
MDL Number: MFCD00006235
CAS Number: 6636-78-8
EINECS: 229-635-3

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR