2,6-Dichloro-3-nitropyridine ≥96% for synthesis

Supplier: Matrix Scientific

011128-25G 011128-100G
101799-946EA 23.08 USD
101799-946 101799-948
2,6-Dichloro-3-nitropyridine ≥96% for synthesis
2,6-Dichloro-3-nitropyridine

Hazard code(s): Irritant

Formula: C₅H₂Cl₂N₂O₂
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00006234
CAS Number: 16013-85-7
EINECS: 240-151-1

Order Now

Specification Test Results

Melting point 57-59°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR