2-Fluoro-3-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

011123-1G 011123-5G 011123-25G
101799-932EA 15.38 USD
101799-932 101799-934 101799-936
2-Fluoro-3-(trifluoromethyl)pyridine ≥98%
2-Fluoro-3-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₃F₄N
MW: 165.09 g/mol
MDL Number: MFCD04973416
CAS Number: 65753-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR