4-Chloro-3-nitropyridine ≥90%, tech.
Supplier: Matrix Scientific
011116-5G
011116-25G
101799-902EA
46.15
USD
101799-902
101799-904
4-Chloro-3-nitropyridine ≥90%, tech.
4-Chloro-3-nitropyridine
Hazard code(s): Irritant, Store in Freezer
Formula:
C₅H₃ClN₂O₂ Melting Pt: ∼32 °C |
MDL Number:
MFCD02683025 CAS Number: 13091-23-1 |
Specification Test Results
Melting point | ca 32°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...