2,4-Dihydroxypyridine ≥98%
Supplier: Matrix Scientific
|
Warning
|
011108-5G
011108-25G
101799-868EA
52.31
USD
101799-868
101799-870
2,4-Dihydroxypyridine ≥98%
2,4-Dihydroxypyridine
Hazard code(s): Irritant
Formula:
C₅H₅NO₂ Boiling Pt: 315 °C (1013 hPa) Melting Pt: ∼267 °C (dec.) |
MDL Number:
MFCD00006273 CAS Number: 626-03-9 EINECS: 210-924-8 |
Specification Test Results
Melting point | ca 267°C(de |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...