2-Methoxynicotinic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid

011102-25G 011102-100G
101799-858EA 29.23 USD
101799-858 102663-582
2-Methoxynicotinic acid ≥98% for synthesis
2-Methoxynicotinic acid

Hazard code(s): Irritant

Formula: C₇H₇NO₃
Melting Pt: 147…149 °C
MDL Number: MFCD00085110
CAS Number: 16498-81-0

Order Now

Specification Test Results

Melting point 147-149°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR