Ethyl-2-chloronicotinate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate

011101-25G 011101-100G
101799-852EA 29.23 USD
101799-852 101799-854
Ethyl-2-chloronicotinate ≥98%
Ethyl-2-chloronicotinate

Hazard code(s): Irritant

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD00674129
CAS Number: 1452-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR