2-Chloroisonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-pyridinecarboxylic acid, 2-Chloropyridine-4-carboxylic acid

011096-25G 011096-100G 011096-500G
101799-842EA 29.23 USD
101799-842 101799-844 101799-846
2-Chloroisonicotinic acid ≥98%
2-Chloroisonicotinic acid

Hazard code(s): Irritant

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: 230…232 °C
MDL Number: MFCD00191402
CAS Number: 6313-54-8

Order Now

Specification Test Results

Melting point 230-232°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR