2-Fluoro-5-methylpyridine ≥99%

Supplier: Matrix Scientific
Danger

Synonyms: 2-Fluoro-5-picoline

011093-25G 011093-100G
101799-834EA 21.54 USD
101799-834 101799-836
2-Fluoro-5-methylpyridine ≥99%
2-Fluoro-5-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 153…155 °C
Density: 1.077 g/cm³ (20 °C)
MDL Number: MFCD00041225
CAS Number: 2369-19-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 153-155°C
Density 1.077
ND20 1.471

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR