2-Amino-5-bromo-4-methylpyridine ≥97%

Supplier: Matrix Scientific
Warning

011092-25G 011092-100G
101799-830EA 40 USD
101799-830 101799-832
2-Amino-5-bromo-4-methylpyridine ≥97%
2-Amino-5-bromo-4-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD03427660
CAS Number: 98198-48-2

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR