Indole-2-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 1H-Indole-2-carboxylic acid

011089-25G 011089-100G
101799-816EA 21.54 USD
101799-816 101799-818
Indole-2-carboxylic acid ≥98%
Indole-2-carboxylic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 203 - 206 °C
MDL Number: MFCD00005611
CAS Number: 1477-50-5
EINECS: 216-030-4

Order Now

Specification Test Results

Melting point 203-206°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR