Indole-5-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 1H-Indole-5-carboxylic acid

011088-1G 011088-5G
101799-812EA 20 USD
101799-812 101799-814
Indole-5-carboxylic acid ≥98%
Indole-5-carboxylic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 211 - 213 °C
Storage Temperature: Freezer
MDL Number: MFCD00005678
CAS Number: 1670-81-1
EINECS: 216-799-6

Order Now

Specification Test Results

Melting point 212-213°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR