2-Bromo-4-methylpyridine ≥97%
Supplier: Matrix Scientific
011081-25G
011081-100G
101799-802EA
30.77
USD
101799-802
102832-826
2-Bromo-4-methylpyridine ≥97%
2-Bromo-4-methylpyridine
Hazard code(s): Irritant
Formula:
C₆H₆BrN Boiling Pt: 87 °C (10mm) Melting Pt: 218…228 °C Density: 1.545 g/cm³ (20 °C) |
MDL Number:
MFCD00082590 CAS Number: 4926-28-7 |
Specification Test Results
Boiling point | 87°C/10mm |
Density | 1.545 |
ND20 | 1.561 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...