2-Bromoisonicotinic acid ≥98%
Supplier: Matrix Scientific
011080-25G
011080-100G
101799-796EA
43.08
USD
101799-796
101799-798
2-Bromoisonicotinic acid ≥98%
2-Bromoisonicotinic acid
Hazard code(s): Irritant
Formula:
C₆H₄BrNO₂ Melting Pt: 229…231 °C |
MDL Number:
MFCD01646069 CAS Number: 66572-56-3 |
Specification Test Results
Melting point | 229-231°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...