2-Bromo-6-methylpyridine ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 6-Bromo-2-picoline, 2-Bromo-6-picoline

011079-25G 011079-100G
101799-792EA 53.85 USD
101799-792 101799-794
2-Bromo-6-methylpyridine ≥98% for synthesis
2-Bromo-6-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 102…103 °C (20mm)
Density: 1.512 g/cm³
MDL Number: MFCD00040743
CAS Number: 5315-25-3
EINECS: 226-173-4

Order Now

Specification Test Results

Boiling point 102-103°C/2
Density 1.512
ND20 1.562


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR