5-Aminonicotinic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Aminopyridine-3-carboxylic acid, 5-Amino-3-pyridinecarboxylic acid
010215-5G
010215-25G
101798-100EA
21.54
USD
101798-100
101798-102
5-Aminonicotinic acid ≥97%
5-Aminonicotinic acid
Hazard code(s): Irritant
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol |
MDL Number:
MFCD00129116 CAS Number: 24242-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...