6-Quinolinecarboxylic acid

Supplier: Matrix Scientific
Warning

010038-5G 010038-25G
101797-750EA 44.62 USD
101797-750 101797-752
6-Quinolinecarboxylic acid
6-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 294…295 °C
MDL Number: MFCD00047613
CAS Number: 10349-57-2
EINECS: 233-761-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR