5-Bromo-o-anisidine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-anisidine, 5-Bromo-2-methoxyaniline, 2-Amino-4-bromoanisole

009827-5G 009827-25G
101797-338EA 96.92 USD
101797-338 101797-340
5-Bromo-o-anisidine ≥98%
5-Bromo-o-anisidine

Hazard code(s): Irritant

Formula: C₇H₈BrNO
MW: 202.05 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD04037882
CAS Number: 6358-77-6
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Melting point 94-96°C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR