4-Nitrophenylboronic acid pinacol ester ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane, 4-Nitrophenylboronic acid Pinacol ester, 1-Nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 4-Nitrobenzeneboronic acid pinacol ester

009802-1G 009802-5G
101797-296EA 36.92 USD
101797-296 101797-298
4-Nitrophenylboronic acid pinacol ester ≥95% for synthesis
4-Nitrophenylboronic acid pinacol ester

Hazard code(s): Irritant

Formula: C₁₂H₁₆BNO₄
MDL Number: MFCD02179437
CAS Number: 171364-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR