2-Fluoro-5-pyridineboronic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 6-Fluoropyridine-3-boronic acid, 2-Fluoropyridine-5-boronic acid, 2-Fluoro-5-pyridylboronic acid, 2-Fluoro-5-pyridinylboronic acid, 6-Fluoro-3-pyridinylboronic acid

009786-5G 009786-25G
101797-256EA 60 USD
101797-256 101797-258
2-Fluoro-5-pyridineboronic acid ≥98% for synthesis
2-Fluoro-5-pyridineboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₅H₅BFNO₂
MDL Number: MFCD03411559
CAS Number: 351019-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR