2,3-Dichlorophenylboronic acid ≥98%

Supplier: Matrix Scientific

009778-5G 009778-25G
101797-238EA 64.62 USD
101797-238 101797-240
2,3-Dichlorophenylboronic acid ≥98%
2,3-Dichlorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 242…244 °C
MDL Number: MFCD01075703
CAS Number: 151169-74-3

Order Now

Specification Test Results

Melting point 242-244°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR