2-Chloro-5-pyridineboronic acid ≥96%
Supplier: Matrix Scientific
Synonyms:
2-Chloropyridine-5-boronic acid, 2-Chloro-5-pyridinylboronic acid, 6-Chloro-3-pyridinylboronic acid, (6-Chloropyridin-3-yl)boronic acid, 6-Chloropyridine-3-boronic acid
009769-1G
009769-5G
101797-214EA
15.38
USD
101797-214
101797-216
2-Chloro-5-pyridineboronic acid ≥96%
2-Chloro-5-pyridineboronic acid
Hazard code(s): Irritant, Refrigerate
Formula:
C₅H₅BClNO₂ MW: 157.36 g/mol |
MDL Number:
MFCD03094998 CAS Number: 444120-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...