2-Chloro-4-pyridineboronic acid ≥96%

Supplier: Matrix Scientific

Synonyms: (2-Chloropyridin-4-yl)boronic acid, 2-Chloro-4-pyridinylboronic acid, 2-Chloropyridine-4-boronic acid

009768-1G 009768-5G 009768-25G
101797-208EA 29.23 USD
101797-208 101797-210 101797-212
2-Chloro-4-pyridineboronic acid ≥96%
2-Chloro-4-pyridineboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD03788416
CAS Number: 458532-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR