2-Bromo-5-pyridineboronic acid ≥96%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-3-pyridinylboronic acid, 2-Bromopyridine-5-boronic acid, (6-Bromopyridin-3-yl)boronic acid, 2-Bromo-5-pyridinylboronic acid, 6-Bromopyridine-3-boronic acid

009762-1G 009762-5G
101797-192EA 16.92 USD
101797-192 101797-194
2-Bromo-5-pyridineboronic acid ≥96%
2-Bromo-5-pyridineboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₅H₅BBrNO₂
MW: 201.81 g/mol
MDL Number: MFCD03411558
CAS Number: 223463-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR