(4-Chloro-2-fluorophenyl)acetic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
4-Chloro-2-fluorophenylacetic acid
009621-5G
009621-25G
101797-158EA
32.31
USD
101797-158
101797-160
(4-Chloro-2-fluorophenyl)acetic acid ≥98%
(4-Chloro-2-fluorophenyl)acetic acid
Hazard code(s): Irritant
Formula:
(C₆H₃ClF)CH₂COOH MW: 188.59 g/mol |
MDL Number:
MFCD03095402 CAS Number: 194240-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...