4-Chloro-2-fluorobenzyl alcohol

Supplier: Matrix Scientific

009618-5G 009618-25G
101797-154EA 53.85 USD
101797-154 101797-156
4-Chloro-2-fluorobenzyl alcohol
4-Chloro-2-fluorobenzyl alcohol
Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD00143285
CAS Number: 56456-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR