2-Chloro-4-fluoroanisole

Supplier: Matrix Scientific
Warning

009610-1G 009610-5G
101797-150EA 290.77 USD
101797-150 101797-152
2-Chloro-4-fluoroanisole
2-Chloro-4-fluoroanisole

Hazard code(s): Irritant

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD00672965
CAS Number: 2267-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR