2-Fluoro-p-toluic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
009509-5G
009509-25G
101797-138EA
64.62
USD
101797-138
101797-140
2-Fluoro-p-toluic acid ≥98%
2-Fluoro-p-toluic acid
Hazard code(s): Irritant
Formula:
C₈H₇FO₂ MW: 154.14 Melting Pt: 186…189 °C |
MDL Number:
MFCD03092906 CAS Number: 7697-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...