4-Chloro-3-fluorobenzyl alcohol ≥98%

Supplier: Matrix Scientific

009485-1G 009485-5G 009485-25G
101797-124EA 20 USD
101797-124 101797-126 101797-128
4-Chloro-3-fluorobenzyl alcohol ≥98%
4-Chloro-3-fluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD00143292
CAS Number: 202925-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR