2-Bromo-4-fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

009433-5G 009433-25G
101797-098EA 15.38 USD
101797-098 101797-100
2-Bromo-4-fluorobenzonitrile ≥98%
2-Bromo-4-fluorobenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₇H₃BrFN
MDL Number: MFCD00672924
CAS Number: 36282-26-5
EINECS: 252-949-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR