5-Bromo-2-chlorobenzoic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
009432-100G
009432-500G
101797-094EA
67.69
USD
101797-094
101797-096
5-Bromo-2-chlorobenzoic acid ≥98%
5-Bromo-2-chlorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄BrClO₂ MW: 235.46 g/mol Boiling Pt: 316 °C (1013 hPa) Melting Pt: 157 - 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002415 CAS Number: 21739-92-4 EINECS: 244-558-5 |
Specification Test Results
Melting point | 159-160°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...