(3-Fluoro-4-methoxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-methoxyphenylacetic acid

009245-5G 009245-25G 009245-100G
101796-708EA 23.08 USD
101796-708 101796-710 101796-712
(3-Fluoro-4-methoxyphenyl)acetic acid ≥98%
(3-Fluoro-4-methoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: C₉H₉FO₃
MW: 184.17 g/mol
MDL Number: MFCD00060353
CAS Number: 452-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR