4-Bromo-2-nitrobenzoic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
009241-5G
009241-25G
009241-100G
101796-690EA
23.08
USD
101796-690
101796-692
101796-694
4-Bromo-2-nitrobenzoic acid ≥97%
4-Bromo-2-nitrobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄BrNO₄ Melting Pt: 165…169 °C |
MDL Number:
MFCD01013599 CAS Number: 99277-71-1 |
Specification Test Results
Melting point | 165-169 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...