2-Fluoro-5-hydroxybenzoic acid

Supplier: Matrix Scientific

009240-5G 009240-25G
101796-686EA 118.46 USD
101796-686 101796-688
2-Fluoro-5-hydroxybenzoic acid
2-Fluoro-5-hydroxybenzoic acid
Formula: C₇H₅FO₃
MDL Number: MFCD01862084
CAS Number: 51446-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR