7-Benzyloxyindole ≥98%

Supplier: Matrix Scientific

Synonyms: 7-(Benzyloxy)-1H-indole

009237-1G 009237-5G 009237-25G
101796-678EA 27.69 USD
101796-678 101796-680 101796-682
7-Benzyloxyindole ≥98%
7-Benzyloxyindole

Hazard code(s): Irritant, Store in Freezer

Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
MDL Number: MFCD00037974
CAS Number: 20289-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR