2-Chloronicotinic acid ≥99%

Supplier: Matrix Scientific
Warning

009230-100G 009230-250G
101796-670EA 73.85 USD
101796-670 101796-672
2-Chloronicotinic acid ≥99%
2-Chloronicotinic acid

Hazard code(s): Irritant

Formula: C₆H₄ClNO₂
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: ∼177 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006236
CAS Number: 2942-59-8
EINECS: 220-937-0

Order Now

Specification Test Results

Melting point ca 177°C(de


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR