1-Bromo-3-chloro-2-fluorobenzene ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Chloro-2-fluorobromobenzene
009229-5G
009229-25G
101796-666EA
15.38
USD
101796-666
101796-668
1-Bromo-3-chloro-2-fluorobenzene ≥95%
1-Bromo-3-chloro-2-fluorobenzene
Hazard code(s): Irritant
Formula:
C₆H₃BrClF MW: 209.44 g/mol |
MDL Number:
MFCD03412182 CAS Number: 144584-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...