4'-(Trifluoromethyl)biphenyl-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)-2'-biphenylcarboxylic acid, 4'-(Trifluoromethyl)-2-biphenylcarboxylic acid, 4'-Trifluoromethyl-2-biphenyl carboxylic acid, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid

009201-1G 009201-5G
101796-628EA 15.38 USD
101796-628 101796-630
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid ≥97%
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid

Hazard code(s): Irritant

Formula: C₁₄H₉F₃O₂
MW: 266.22 g/mol
Melting Pt: 169…171 °C
MDL Number: MFCD00075353
CAS Number: 84392-17-6

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR