5-Benzyloxyindole

Supplier: Matrix Scientific

Synonyms: 5-Benzyloxy-1H-indole, 5-(Benzyloxy)-1H-indole

009194-5G 009194-25G
101796-610EA 15.38 USD
101796-610 101796-612
5-Benzyloxyindole
5-Benzyloxyindole

Hazard code(s): Irritant, Store in Freezer

Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Boiling Pt: 165 °C (0.1 -rr)
Melting Pt: 107 °C
MDL Number: MFCD00005676
CAS Number: 1215-59-4
EINECS: 214-930-1

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR