2-Chloro-3-fluoropyridine ≥97%

Supplier: Matrix Scientific

009190-5G 009190-25G 009190-100G
101796-604EA 15.38 USD
101796-604 101796-606 101796-608
2-Chloro-3-fluoropyridine ≥97%
2-Chloro-3-fluoropyridine

Hazard code(s): Irritant

Formula: C₅H₃ClFN
MW: 131.54 g/mol
Boiling Pt: 64…65 °C (80mm)
Density: 1.323 g/cm³
MDL Number: MFCD03095302
CAS Number: 17282-04-1
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Boiling point 64-65°C/80m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR