1,1,1,3-Tetrachlorotetrafluoropropane ≥97%

Supplier: Matrix Scientific

009159-5G 009159-25G
101796-532EA 158.46 USD
101796-532 101796-534
1,1,1,3-Tetrachlorotetrafluoropropane ≥97%
1,1,1,3-Tetrachlorotetrafluoropropane

Hazard code(s): Ozone Depleter

Formula: Cl₃CCF₂CClF₂
Boiling Pt: 114 °C
Melting Pt: –92.8 °C
Density: 1.6927 g/cm³
MDL Number: MFCD00039325
CAS Number: 2268-46-4
EINECS: 218-868-6

Order Now

Specification Test Results

Boiling point 114°C
Melting point -92.8°C
Density 1.6927
ND20 1.3994

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR