Potassium heptadecafluorooctane-1-sulphonate ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: Potassium perfluorooctanesulphonate, Heptadecafluoro-1-octanesulphonic acid potassium salt, Potassium perfluorooctane-1-sulphonate, Perfluoro-1-octanesulphonic acid potassium salt, Perfluorooctanesulphonic acid, potassium salt, Heptadecafluorooctanesulphonic acid potassium salt

009151-5G 009151-25G
101796-504EA 163.46 USD
101796-504 101796-506
Potassium heptadecafluorooctane-1-sulphonate ≥98% for synthesis
Potassium heptadecafluorooctane-1-sulphonate

Hazard code(s): Irritant

Formula: C₈F₁₇KO₃S
MW: 538.22
Melting Pt: 285 °C
MDL Number: MFCD00066407
CAS Number: 2795-39-3
EINECS: 220-527-1

Order Now

Specification Test Results

Melting point 285°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR