1H,1H-Hentriacontafluoro-1-hexadecanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-Hentriacontafluoro-1-hexadecanol, 1H,1H-Perfluoro-1-hexadecanol

009106-1G 009106-5G
101796-360EA 56.92 USD
101796-360 101796-362
1H,1H-Hentriacontafluoro-1-hexadecanol ≥95%
1H,1H-Hentriacontafluoro-1-hexadecanol

Hazard code(s): Irritant

Formula: C₁₆H₃F₃₁O
MDL Number: MFCD00236626
CAS Number: 216144-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR