1H,1H,2H,2H-Heptadecafluoro-1-decanol ≥97%
Supplier: Matrix Scientific
Synonyms:
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanol, 1H,1H,2H,2H-Perfluoro-1-decanol, 1H,1H,2H,2H-Perfluorodecan-1-ol
009095-25G
009095-100G
101796-328EA
58.46
USD
101796-328
101796-330
1H,1H,2H,2H-Heptadecafluoro-1-decanol ≥97%
1H,1H,2H,2H-Heptadecafluoro-1-decanol
Hazard code(s): Irritant
Formula:
C₁₀H₅F₁₇O MW: 464.12 g/mol Boiling Pt: 112…114 °C (10mm) Melting Pt: 50 °C |
MDL Number:
MFCD00039544 CAS Number: 678-39-7 EINECS: 211-648-0 |
Specification Test Results
Boiling point | 112-114°C/1 |
Melting point | 50°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...