4-Nitroindole ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Nitro-1H-indole

009071-1G 009071-5G 009071-25G
101796-272EA 15.38 USD
101796-272 101796-274 102832-802
4-Nitroindole ≥98%
4-Nitroindole

Hazard code(s): Irritant, Irritant-Harmful, Store in Freezer

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 206 - 209 °C
MDL Number: MFCD00010056
CAS Number: 4769-97-5

Order Now

Specification Test Results

Melting point 205-207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR