Bismuth(III) trifluoromethanesulphonate for synthesis
Supplier: Matrix Scientific
Synonyms:
Bismuth triflate, Bismuth(III) trifluoromethanesulphonate
009065-25G
009065-100G
101796-250EA
223.08
USD
101796-250
101796-252
Bismuth(III) trifluoromethanesulphonate for synthesis
Bismuth(III) trifluoromethanesulphonate
Formula:
C₃BiF₉O₉S₃ |
MDL Number:
MFCD02093669 CAS Number: 88189-03-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...