4-Nitro-2-(trifluoromethyl)anisole ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Methoxy-4-nitro-2-(trifluoromethyl)benzene, 2-Methoxy-5-nitrobenzotrifluoride
009020-10G
009020-25G
101796-134EA
23.08
USD
101796-134
101796-136
4-Nitro-2-(trifluoromethyl)anisole ≥98%
4-Nitro-2-(trifluoromethyl)anisole
Hazard code(s): Irritant
Formula:
C₈H₆F₃NO₃ MW: 221.14 g/mol Melting Pt: 79…81 °C |
MDL Number:
MFCD00007328 CAS Number: 654-76-2 EINECS: 211-507-3 UN: 2811 ADR: 6.1,I |
Specification Test Results
Melting point | 79-81°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...